Documenti fondamentali
Z173134
Sigma-Aldrich
Aldrich® Schlenk-type adapter
90°-Bent adapter with glass stopcock and hose barb, male joint: threaded 14/20
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.43
Giunto
male joint: threaded14/20
Descrizione generale
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)