Key Documents
Z185221
Double-tipped needle
L 24 in., gauge 14
About This Item
Codice UNSPSC:
41122002
NACRES:
NB.03
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Cadence Science
Lunghezza
24 in.
Gauge
14
Descrizione generale
304 SS with deflected tips.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 5
1 of 5
Double-ended needle gauge 16, L 36 in.
Z102458
Double-ended needle
Double-ended needle gauge 16, L 24 in.
Z102431
Double-ended needle
Dissolution cannulae gauge 14, 316 stainless steel hub, L 7 1/2 in.
CAD9881
Dissolution cannulae
Dissolution cannulae gauge 14, 316 stainless steel hub, L 4 3/8 in.
CAD9880
Dissolution cannulae