Z154407
Ace Trubore® PTFE 4-piece stirrer bearing
joint: ST/NS 24/40, fits shaft o.d., 10 mm
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.72
Prodotti consigliati
Giunto
joint: ST/NS24/40
Caratteristiche
fits shaft o.d. 10 mm
Produttore/marchio commerciale
Ace Glass 806643
Categorie correlate
Descrizione generale
Self-aligning, lubricant-free PTFE Trubore bearing for use with SS or polished glass stirring shafts at approximately 200rpm. Consists of PTFE inner Trubore bearing, FETFE® compression saddle O-ring, glass-reinforced bushing and lock-nut with FETFE O-ring, and threaded glass adapter. The lock permits stirring in either direction without fear of unthreading. The compression saddle O-ring maintains constant force with little attention. Vacuum to 1 mm Hg. Not recommended for use with ground-glass shafts.
Note legali
FETFE is a registered trademark of Ace Glass, Inc.
Trubore is a registered trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z154407-1EA | 4061832347707 |