Documenti fondamentali
Z504742
Ace O-rings, FETFE®
I.D. 12.4 mm, wall size 1.78 mm
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Confezionamento
pkg of 12 ea
Produttore/marchio commerciale
Ace Glass 7855712
Diam. int.
12.4 mm
Misura parete
1.78 mm
Note legali
FETFE is a registered trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 5
1 of 5
Ace pressure tube bushing type, Front seal, capacity 60 mL, L × O.D. 10.2 cm × 38.1 mm
Z568767
Ace pressure tube
Ace pressure tube bushing type, Back seal, capacity ~35 mL, L × O.D. 17.8 cm × 25.4 mm
Z181072
Ace pressure tube
Ace pressure tube bushing type, Front seal, capacity ~35 mL, L × O.D. 17.8 cm × 25.4 mm
Z181102
Ace pressure tube
Ace pressure tube bushing type, Front seal, capacity ~21 mL, L × O.D. 20.3 cm × 19 mm
Z564621
Ace pressure tube
Ace pressure tube bushing type, Front seal, capacity ~4 mL, L × O.D. 10.2 cm × 13 mm
Z564559
Ace pressure tube
Global Trade Item Number
SKUGTIN
Z504742-1PAK4061832363455