Z125997
Ace Trubore® 3-piece glass stirrer bearing
joint: ST/NS 24/40, fits shaft o.d., 10 mm
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.72
Prodotti consigliati
Giunto
joint: ST/NS24/40
Caratteristiche
fits shaft o.d. 10 mm
Produttore/marchio commerciale
Ace Glass 8042115
Categorie correlate
Descrizione generale
Self-aligning, three-piece bearing consisting of a glass inner Trubore bearing, nylon bushing with O-ring, and threaded glass adapter. Inner bearing will spin if shaft binds or will self-align at O-ring if motor is slightly cocked. Inner bearing and bushing can be used with any joint size.
Note legali
Trubore is a registered trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z125997-1EA | 4061832347035 |