Z122890
Ampule breaker/collar
For 5-10 mL ampules
About This Item
Codice UNSPSC:
41121807
NACRES:
NB.43
Prodotti consigliati
Descrizione generale
How to Use:
1. Slip polyethylene breaker/ collar over top of ampule.
2. Grip bottom of ampule with one hand and breaker/collar with other.
3. Gently snap top of ampule off.
4. Discard collar and ampule top.
1. Slip polyethylene breaker/ collar over top of ampule.
2. Grip bottom of ampule with one hand and breaker/collar with other.
3. Gently snap top of ampule off.
4. Discard collar and ampule top.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)