Documenti fondamentali
Z114243
Supelco
Hand-operated aluminum cap crimper
O.D. 20 mm
Sinonimo/i:
Manual Cap Crimping Tool
About This Item
Codice UNSPSC:
41122001
NACRES:
NB.11
D.E.
20 mm
Descrizione generale
  • Used to attach aluminum seals to bottles and vials with a crimp/serum finish
  • Cushioned ergonomic handle reduces hand fatigue
  • Labeled for quick size identification
  • Polished crimping jaws provide consistent sealing
  • 2 year warranty
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 7
1 of 7
Crimpers Hand crimper, for use with 11mm crimp seals, pkg of 1 ea
Supelco
33195
Crimpers
Hand crimper, adjustable Hand crimper, adjustable, for use with 11mm crimp top vials, pkg of 1 ea
22313-U
Hand crimper, adjustable
Stoppers stopper diam. 20 mm, gray butyl rubber, pkg of 100 ea
Supelco
27232
Stoppers
Crimpers Hand crimper, for use with 13mm seals, pkg of 1 ea
33279-U
Crimpers