Documenti fondamentali
Z114111
Sigma-Aldrich
Septa for 20 mm crimp seals
tan PTFE/silicone septa, size × total thickness × PTFE thickness 20 mm × 0.130 in. × 0.005 in., for use with tear-off seals
About This Item
Codice UNSPSC:
41122001
NACRES:
NB.11
Materiali
tan PTFE/silicone septa
Misura × spess. totale × spess. PTFE
20 mm × 0.130 in. × 0.005 in.
Descrizione generale
It is used as an air-tight seal, and prevents contamintion.
Note legali
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
Z114111-100EA4061832314112
Z114111-10EA4061832314129