SYND681420
Synthware™ distillation distribution adapter with vacuum hose connection
joint: ST/NS 14/20
Sinonimo/i:
Synthware D681420, radial style distillation receiver
About This Item
Codice UNSPSC:
41104812
NACRES:
NB.51
Prodotti consigliati
Giunto
joint: ST/NS14/20
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware D681420
Descrizione generale
Cow-type design having three receivers spaced 40° apart. 105° angle between top outer joint and lower inner joints.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)