CAD6520
1-way Luer-to-Luer adapter
MLL to MLL (316SS)
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.03
Prodotti consigliati
Confezionamento
pack of 5 ea
Produttore/marchio commerciale
Cadence Science 6520
Categorie correlate
Descrizione generale
- Plated brass or 316 stainless steel
- Meets ISO 594 specifications
- Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Micro-Mate® Female Luer Lock (MMFLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.
Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.
Male Luer (ML) - male Luer slip that fits with a female Luer.
Note legali
Micro-Mate is a registered trademark of Cadence Science
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
CAD6520-5EA | 4061832517872 |