BR144645
BRAND® BISTABIL® ground glass stopper, conical joint
joint: ST/NS 45/40
Sinonimo/i:
BRAND® BISTABIL® ground glass stopper conical joint stopper, Conical joint stoppers, Stoppers
About This Item
Codice UNSPSC:
41122500
NACRES:
NB.51
Prodotti consigliati
Giunto
joint: ST/NS45/40
Descrizione
hexagonal grip
hollow
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 144645
Categorie correlate
Descrizione generale
Conical ground joint stopper made from Duran®. Hollow with drip tip and hexagonal grip. Meets DIN 12 252.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Bistabil is a registered trademark of Lenz GmbH
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)