Z304638
Quickfit® ground-glass stoppers
joint: ST/NS 24/29
Sinonimo/i:
SB24
About This Item
Codice UNSPSC:
41122500
NACRES:
NB.35
Prodotti consigliati
Giunto
joint: ST/NS24/29
Produttore/marchio commerciale
Quickfit SB 24
Disponibilità
not available in USA and Canada
Descrizione generale
- Ideally suited for use with Quickfit® jointedware and other glassware having standard ground glass sockets
- All Quickfit® stoppers conform to the standard tapers as outlined in ISO 383
- Borosilicate glass for chemical durability and high temperature performance
- Hollow-blown construction to reduce weight
- Hexagonal head prevents the stopper from rolling and provides a good grip
- Top of stopper is flat, allowing them to be stood on end and avoid contamination of the ground surface
Note legali
Quickfit is a registered trademark of SciLabware Limited
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z304638-1EA | 4061832830179 |