28403-U
1 of 10
Supelco
28423-U
二噁英样品制备套装
1.19851
Vacuum manifold
28404-U
二噁英空色谱柱
SYNS560008
Synthware™ 180 degree high vacuum valve
28449-U
具有旋阀的顶部烧瓶
Millipore
XX2504700
真空过滤歧管
28398-U
6.35mm/10mm 异径转接头
SYNM490064
Synthware™ vacuum manifold with PTFE stopcocks
24452
Hamilton® Gastight® 数字注射器
SYNM202405
Synthware™ vacuum manifold with high vacuum valves
includes long stem stopcocks, 10 mm O.D.
solid phase extraction (SPE): suitable
food and beverages
分析化学_分析色谱_分析试剂-默克生命科学
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.