H43ESK00P
1 of 10
Millipore
HGTC600T0
Cartridge Housing Sanitary Gaskets 6 in. PTFE encapsulated fluoroelastomer
Supelco
1.08027
pH Test
1.09533
pH指示条pH 5.0-10.0
Z678538
2Mag MIX 1 eco magnetic stirrer
BR705892
BRAND® Transferpette® S Pipette Starter Kit
Sigma-Aldrich
Z516163
Jacketed falling-film distillation head
MABE78
Anti-MiTF Antibody, clone 3D1.2
MAB2642
Anti-Fibrillin-2 Antibody, clone 48
MAB1904
Anti-Laminin-1 A&B chains Antibody, cross region, clone AL-2
C61984
2-氯-3-硝基甲苯
EPDM rubber
100
for use with Series 2000 Sanitary Cartridge Housings/3-tube