Z543373
1 of 10
Millipore
XX1104707
O形圈,(5-329)用于Swinnex 47 mm过滤器支架,硅胶
Sigma-Aldrich
Z250201
Wheaton wide-mouth bottle
Z290432
Sigma-Aldrich 真空过滤组件
Z504343
Ace O-rings, silicone
SX0004701
Base O-ring for Swinnex 47 mm filter holder, silicone
F2036
Nalgene® filtration assembly for 47 mm filter
XX2504754
Funnel Cover, 47mm, silicone
XX11J4750
Sterifil换膜过滤器,500 mL
Z325155
BRAND® BLAUBRAND® ETERNA volumetric pipette, calibrated To Deliver
Z156485
O 型圈套装
pkg of 6 ea
IR spectroscopy: suitable