主要文件
MT51343153
Mettler-Toledo InLab® Solids & Solids Pro pH electrodes
S7 connector
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41115604
NACRES:
NB.79
材料
glass membrane (LoT)
glass shaft
描述
Reference electrolyte XEROLYT EXTRA Polymer
Reference system ARGENTHAL
without cable
製造商/商標名
Mettler-Toledo 51343153
溫度測量範圍
0-80 °C
長度 × 直徑
25 mm × 6 mm
pH值範圍
1-11
正在寻找类似产品? 访问 产品对比指南
一般說明
InLab® Solids & Solids Pro are combination pH electrodes with option of a variety of connectors.
The spear shaped tip has been designed for easy piercing of fruit, cheese, meats, etc
Designed for measuring samples with a tough exterior, the InLab® Solids has a "spear" shaped bulb for easy piercing.
The InLab® Solids electrode has been designed with long life in mind. The fine-pored ceramic junction used in conventional electrodes can easily become contaminated when measuring the pH of food samples. In contrast the InLab® Solids electrode with a solid polymeric electrolyte requires no junction at all. As a result these sensors are particularly insensitive to contamination. The reference electrode on these sensors is in direct contact with the measured medium by means of an open junction, hence there is no junction to block.
Service life and measurement accuracy are further improved by the ARGENTHAL reference system which reduces loss of silver chloride from the reference wire at differing sample temperatures.
The tough glass shaft resists breakage and makes the InLab® Solids suitable for near process and factory applications.
  • "Spear" shaped tip
  • XEROLYT PLUS Polymer electrolyte
  • ARGENTHAL reference
  • Detachable cable
  • Rugged glass shaft
法律資訊
ARGENTHAL is a trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
XEROLYT is a trademark of Mettler-Toledo, Inc.
历史批次信息供参考:
分析证书(COA)