Documenti fondamentali
57172
Supelco
ENVI-8 DSK SPE Disk
diam. 47 mm, pk of 24
About This Item
Codice UNSPSC:
41115712
Confezionamento
pk of 24
Livello qualitativo
tecniche
solid phase extraction (SPE): suitable
Diametro
47 mm
applicazioni
food and beverages
Tecnica di separazione
reversed phase
Descrizione generale
Retention Mechanism: Reversed-phase
Sample Matrix Compatibility: Aqueous solutions (drinking water)
  • The SPE membrane equivalents of ENVI-18 and ENVI-8 packed bed SPE sorbents
  • Porous glass fiber membranes embedded with C18 or C8 modified silica particles.
  • Provides faster flow rates and exhibits less clogging than PTFE discs for the extraction of organic contaminants from drinking water samples
  • Typical applications include polynuclear aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), phthalates, semivolatile organics, paraquat and diquat, pesticides and herbicides
Note legali
ENVI is a trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Articoli
Solid Phase Extraction: Reversed-Phase Methodology
Protocolli
HPLC Analysis of Paraquat and Diquat on SUPELCOSIL LC-18 after SPE using ENVI-8 DSK
Chromatograms
application for SPE, application for HPLC