Documenti fondamentali
502375
Micro-Metering Valve (Low Pressure)
10-32 fingertight fittings
About This Item
Codice UNSPSC:
41115854
eCl@ss:
32060101
Materiali
PEEK valve
Descrizione
micro-metering low pressure
Produttore/marchio commerciale
Upchurch Scientific, Inc P-446
Parametri
800 psi max. pressure
Raccordi
10-32 fingertight fittings
Applicazioni
Micro-Metering valve (low pressure) can be used as a component of supercritical fluid extraction apparatus employing supercritical carbon dioxide, which may be used in the extraction and refining of soya-bean oil from full fat soya bean flakes and essential oil from the roots of Vetiveria zizanioides.
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Response surface method applied to supercritical carbon dioxide extraction of Vetiveria zizanioides essential oil.
Danh T L, et al.
Chemical Engineering Journal, 155(3), 617-626 (2009)
Characterization of soybean oil extracted by supercritical carbon dioxide and hexane.
Friedrich P J and List R G.
Journal of Agricultural and Food Chemistry, 30(1), 192-193 (1982)