Documenti fondamentali
Z719420
Sorenson low binding aerosol barrier tips
MultiGuard, maximum volume 100 μL
Sinonimo/i:
aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, low bind tip, pipet tip
About This Item
Codice UNSPSC:
41121601
NACRES:
NB.01
Materiali
colorless
polypropylene
Sterilità
sterile
Caratteristiche
DNase free
RNase free
barrier
Confezionamento
pack of 96 ea (Racked)
Produttore/marchio commerciale
Sorenson 36060T
Volume massimo
100 μL
Volume
1—100 μL
Tipo di binding
low binding surface
Descrizione generale
Tips feature a new surface technology designed for quantitative assays in which sample binding can significantly alter results or for critical small volume protocols that require a high degree of accuracy. Ideal for molecular biology applications, such as PCR. Inert hydrophobic barrier prevents pipettor contamination.
Caratteristiche e vantaggi
  • Suitable for DNA amplification and radioisotope handling
  • Over-pipetting does not contaminate valuable samples
  • Universal fit on research-grade pipettors
  • Filter devoid of leachable additives
  • Small micron pore sizes, <20 m
  • Hydrophobic surface to lower surface tension
  • Minimal sample binding
  • Enhanced accuracy and precision
  • Additives-free such as silicone
  • Eliminates leaching and subsequent sample degradation
Note legali
Sorenson is a trademark of Sorenson BioScience, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)