Z719439
Sorenson™ low binding aerosol barrier tips
MultiGuard, no graduation, maximum volume 200 μL
Sinonimo/i:
Aerosol Barrier Tips
About This Item
Codice UNSPSC:
41121601
NACRES:
NB.01
Prodotti consigliati
Materiali
colorless
polypropylene
Sterilità
sterile
sterile
Caratteristiche
DNase free
RNase free
barrier
Confezionamento
pack of 96 ea (Racked)
Produttore/marchio commerciale
Sorenson 14220T
Volume massimo
200 μL
Volume
200 μL
Tipo di binding
low binding surface
Descrizione generale
Tips feature a new surface technology designed for quantitative assays in which sample binding can significantly alter results or for critical small volume protocols that require a high degree of accuracy. Ideal for molecular biology applications, such as PCR. Inert hydrophobic barrier prevents pipettor contamination.
Note legali
Sorenson is a trademark of Sorenson BioScience, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)