Key Documents
Z601195
Hettich® MIKRO 120 centrifuge accessories
adapter for 0.5-0.8 mL tubes 8x45 mm
About This Item
Codice UNSPSC:
41103911
NACRES:
NB.22
Caratteristiche
Holds 0.5 - 0.8 mL tubes
Confezionamento
pack of 24 ea
Produttore/marchio commerciale
Hettich 2023
tecniche
centrifugation: suitable
Compatibilità
for use with 1213-A, 1226-A, 1252-A, 1258-A, 2428, 2434, 2437 (rotors)
Note legali
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG