TMO1201-0030
Nalgene® Griffin beakers
polypropylene, volume 30 mL, case of 18 ea
Sinonimo/i:
PP beaker, nalgene PP beaker
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Materiali
polypropylene
Confezionamento
case of 18 ea
Produttore/marchio commerciale
Nalgene 1201-0030
Volume
30 mL
Descrizione generale
Low form. With silk-screened graduations, Nalgene catalog number, size and resin codes, max. use temp., and "no flame" symbol. Meet the ISO/DIS 7056 International Standards for Laboratory Plasticware Accuracy. Chemically resistant. Autoclavable. Not for use on a hot plate.
Altre note
Browse all plastic beakers
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)