Documenti fondamentali
EP022822233
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-50-8-16, non aerosol tight, (8 x 8- 12 mL round-bottom tubes)
About This Item
Codice UNSPSC:
41103909
Caratteristiche
to hold 8 x 16x105 - 120 mm tubes
to hold 8 x 8 - 12 mL round-bottom tubes
fixed angle rotor
non aerosol tight
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Eppendorf® 022822233 (NA)
tecniche
centrifugation: suitable
Capacità
(8 x 8- 12 mL round-bottom tubes)
Compatibilità
for use with centrifuge models Vacufuge Plus Concentrator
Descrizione generale
Selection of rotors for the Eppendorf Vacufuge Plus.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG