Documenti fondamentali
EP0030128931
Eppendorf® Twin tec PCR plates, 384 wells
colorless plate, conical bottom colorless wells, pkg of 300 ea
About This Item
Codice UNSPSC:
41106300
NACRES:
NB.15
Materiali:
colorless plate
conical bottom colorless wells
Misura:
384 wells
Volume massimo del pozzetto:
40 μm
Materiali
colorless plate
conical bottom colorless wells
Confezionamento
pkg of 300 ea
Produttore/marchio commerciale
Eppendorf® 0030128931
tecniche
PCR: suitable
Misura
384 wells
Volume massimo del pozzetto
40 μm
Descrizione generale
The Eppendorf twin.tec® PCR plate combines extremely thin-walled polypropylene wells for optimal heat transfer to the sample, with robust polycarbonate frames for high rigidity. With raised well rims, twin.tec® plates enable highly effective sealing to reduce the risk of cross-contamination. With twin.tec® PCR plates you’ll have maximum confidence in your results.
Applicazioni
Eppendorf® Twin tec PCR plates, 384 wells are suitable for use in:
  • Standard PCR
  • Real-time PCR
  • High throughput PCR
  • Small volume sample handling
Caratteristiche e vantaggi
  • Batch -tested and certified to be DNA, DNase, RNase, and PCR inhibitors free (PCR clean).
  • Specially optimized rim is suitable for barcodes and automated work processes.
  • The plates are stackable.
  • They adhere to the recommended SBS footprint standards.
  • These plates are ideal for a 384-well thermocycler, well-suited for the Eppendorf Mastercycler pro-384.
  • Maximum volume per well is 40 µL.
  • Upon request, the plates can be provided with barcodes, but this option is available only for skirted and semi-skirted variants.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)