CLS7610
Corning® Transtar® liquid handling system
disposable cartridges
Sinonimo/i:
lab equipment accessory
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.11
Prodotti consigliati
Sterilità
sterile
Produttore/marchio commerciale
Corning 7610
Descrizione generale
The Transtar is a portable, autoclavable liquid handling system designed for use with 96 well plates, A sterile 96 tip disposable cartridge, which loads into the Transtar system, enables liquids to be aspirated, transferred and dispensed over a volume range of 25 - 200 μL in 5 μL increments, Transtar accuracy us rated at 5% at all volume levels.
Note legali
Corning is a registered trademark of Corning, Inc.
TRANSTAR-96 is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)