CLS7160
CellSTACK® Chamber, 10 STACK
Sinonimo/i:
cell culture chamber
About This Item
Prodotti consigliati
Sterilità
sterile
Produttore/marchio commerciale
Corning
Disponibilità
available only in China
Categorie correlate
Descrizione generale
The CellSTACK® Chamber is a sterile and nonpyrogenic cell culture vessel. They consist of ten chambers, having a total cell growth area of 6360 cm2.
Applicazioni
CellSTACK® Chambers have been used extensively in large-scale cell culture experiments, aiding in the attachment and growth of diverse kinds of cells.
Caratteristiche e vantaggi
- The TM="CellSTACK"] Chambers possess superior mechanical strength and structural integrity.
- The presence of self-venting caps helps in the gas exchange process.
- These are 100% leak evaluated.
- The CellSTACK Chambers have larger openings with threaded closures, ensuring easy accessibility to cells while maintaining a secure seal to prevent contamination.
- The CellSTACK chamber has a sterility assurance level of 10-6.
- A hydrophilic and negatively charged surface enhances cell growth and attachment.
- A minimum of 95% confluency is necessary for optimum cell growth and attachment
Note legali
CellSTACK is a registered trademark of Corning, Inc.