CLS6795400D
Corning® Digital Hotplates
AC/DC input 120 V AC, US 3-pin plug, L × W 5 in. × 7 in.
Sinonimo/i:
stirrers
About This Item
Codice UNSPSC:
41102406
NACRES:
NB.76
Prodotti consigliati
Materiali
ceramic plate
Sterilità
non-sterile
Confezionamento
case of 1
Produttore/marchio commerciale
Corning 6795-400D
Disponibilità
not available in EU
Parametri
5-550 °C temp. range
Lungh. × largh.
5 in. × 7 in.
Colore
none
Categorie correlate
Descrizione generale
- Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
- Microprocessor maintains consistent and repeatable temperature settings.
- Bright LED HOT TOP icon lights up when top plate temperature is over 60 °C, even when heat control is turned off.
- Separate temperature sensor provides power cut off if unit overheats
- Extremely durable and heat resistant Pyroceram glass-ceramic top
- Small footprint and low profile
- Meets UL and cUL standards
- 2 year warranty
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)