Documenti fondamentali
CLS480129
Corning® LSE single block, for Corning LSE single block digital dry baths
single block, 5 x 50 mL centrifuge tubes, 1/cs
Sinonimo/i:
equipment replacement accessory
About This Item
Codice UNSPSC:
41102404
NACRES:
NB.15
Caratteristiche
Holds 5 x 50 mL tubes
Confezionamento
case of 1
pack of 1
Produttore/marchio commerciale
Corning 480129
Compatibilità
suitable for (use with Corning LSE digital dry bath heater, single block)
Descrizione generale
Corning LSE single blocks are suitable for the following digital dry baths:
CLS6785SB, CLS6786SB, CLS6787SB
Note legali
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
CLS480129-1EA4061825882581