Documenti fondamentali
CLS3956
Corning® 96 well Polypropylene Deep Well Plate
V-bottom clear, polypropylene, bag of 10 ×, sterile, lid: no
Sinonimo/i:
cell culture plate, multi well plates, multiple well plates
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.13
Materiali
V-bottom clear
polypropylene
polypropylene plate
translucent bottom
Sterilità
sterile
Caratteristiche
lid: no
skirt
Produttore/marchio commerciale
Corning 3956
Confezionamento
bag of 10 ×
Misura
96 wells
Volume pozzetto
0.5 mL
Pozzetti
96 wells , storage block
Capacità
0.5 mL
Compatibilità
suitable for (cell cuture)
Tipo di binding
non-treated surface
Descrizione generale
Corning offers a complete polypropylene storage system consisting of plates, blocks, and mats.
All plates and blocks are made from polypropylene for solvent resistance and are ideal for compound library, combinatorial chemistry and genomics applications. The 96 well polypropylene plates are built to the industry standard footprint. The plates are available in round, flat, and V-bottom formats to meet a wide variety of assay and storage needs.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)