Documenti fondamentali
CLS3591
Corning® 96 Well EIA/RIA Assay Microplate
flat bottom clear, polystyrene, pkg of (individually wrapped), non-sterile, lid: no
Sinonimo/i:
96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, EIA/RIA plate, immunoassay plate
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.13
Materiali:
flat bottom clear
polystyrene
polystyrene
round wells
Misura:
96 wells , 96 well plate
Volume pozzetto:
330-360 μL , Note: CLS3366 and CLS3797 have well volume capacities of 330 uL
Volume di lavoro pozzetto:
75-200 μL
Materiali
flat bottom clear
polystyrene
polystyrene
round wells
Sterilità
non-sterile
Caratteristiche
lid: no
skirt
plate format: 96 well standard
well: flat bottom, clear
Confezionamento
pack of 1
case of 50
pkg of (individually wrapped)
Produttore/marchio commerciale
Corning 3591
Misura
96 wells , 96 well plate
Volume pozzetto
330-360 μL , Note: CLS3366 and CLS3797 have well volume capacities of 330 uL
Volume di lavoro pozzetto
75-200 μL
Compatibilità
suitable for (EIA/RIA; immunoassays)
Tipo di binding
non-treated surface
Descrizione generale
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Binding capacity of approximately 250 ng of mouse IgG/cm2
  • EIA/RIA plates are tested on lot-to-lot basis and the certification results for each lot are available upon request
  • Recommended working volume of up to 75 to 200 μL
  • Conforms to standard microplate footprint and dimensions
  • Without lids, lids supplied separately
These 96 well EIA/RIA plates are designed for in vitro diagnostic immunoassays. All EIA/RIA plates are manufactured from special medical-grade polystyrene for uniform binding, high optical clarity and low background absorption. In addition, the Easy-Wash products are part of the Certified Surface Chemistry Programme, which guarantees protein binding performance to be within QA release specifications.
Note legali
Corning is a registered trademark of Corning, Inc.
Easy-Wash is a trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)