Key Documents
CLS356551
Corning® Falcon® Serological Pipettes
capacity 10 mL, individually wrapped, clear-plastic, sterile
Sinonimo/i:
pipet
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.02
Materiali
polystyrene
Sterilità
sterile
Caratteristiche
individually wrapped clear-plastic
subdivision
Produttore/marchio commerciale
Corning 356551
Capacità
10 mL
Applicazioni
Liquid Handling
Note legali
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.