Z742416
PTFE-faced silicone septa for Q-Tube™ and QianCap™
septa 12 mL, max. pressure 180 psi
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Prodotti consigliati
Materiali
PTFE face
white silicone backing
Produttore/marchio commerciale
Q Labtech 100-000825-001
Parametri
180 psi max. pressure
Descrizione generale
This disposable septa are made of tan PTFE face and white silicone backing. It fits on Q-Tube-12 mL with Adapter-180 psi. The tan PTFE liner has excellent chemical resistance and silicon backing has reseal ability. Using other septa may cause explosion.
Note legali
Q-Tube is a trademark of Q Labtech LLC
QianCap is a trademark of Q Labtech LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z742416-100EA | 4061834419600 |