Z740645
Aldrich® Essentials desiccator
I.D. × H 250 mm × 330 mm, with perforated porcelain plate
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.43
Prodotti consigliati
Diam. int. × alt.
250 mm × 330 mm
Descrizione generale
Heavy-wall, molded, neutral glass, desiccators with ground flange top and lid. Lid has knob top to assist in removing lid. Complete with perforated, bottom, porcelain plate. No direct heat or flame can be used.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)