Z682306
Ace lower distribution head with shut-off valve for condenser "CR" assembly
glass, fits Büchi® models R-200/R-205 and R-114/R-144
Sinonimo/i:
Ace lower distribution head with shut-off valve for condenser "CR" assembly
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Materiali
glass
Produttore/marchio commerciale
Büchi equivalent, 40657
Descrizione generale
The lower distribution head for the Ace condenser "CR" assembly, and fits the Büchi® rotary evaporators.
Note legali
Büchi is a registered trademark of Büchi Labortechnik AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)