Z563625
Vacuum seals for Büchi® Rotavapor® evaporators
VS-26, for R210/215, RII and R300
Sinonimo/i:
Evaporator seal
About This Item
Codice UNSPSC:
41104804
NACRES:
NB.43
Prodotti consigliati
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Büchi 11069167
Descrizione generale
The new vacuum seal VS-26 (Part No. 11069167) of the R-300 has completely replaced the well-known WD-26 (048021). Tests have shown that the new VS-26 has improved significantly in terms of tightness and resistance to the WD-26. The seal (PTFE / NBR) is FDA compliant and, in addition to the R-300, is compatible with older models R-210 and R-215. The scope of delivery of the VS-26 includes not only a short installation info but also the necessary adapter.
IMPORTANT:
When installing the VS-26 the order is essential to not damage the seal:
1. Fasten the steam feed-through tube in the drive of the corresponding rotary evaporator ("click" sound)
2. Place the gasket with the adapter over the DDR and press it until the adapter comes loose from the gasket (keep the adapter!)
3. Fasten the radiator as usual.
IMPORTANT:
When installing the VS-26 the order is essential to not damage the seal:
1. Fasten the steam feed-through tube in the drive of the corresponding rotary evaporator ("click" sound)
2. Place the gasket with the adapter over the DDR and press it until the adapter comes loose from the gasket (keep the adapter!)
3. Fasten the radiator as usual.
Note legali
Büchi is a registered trademark of Büchi Labortechnik AG
Rotavapor is a registered trademark of Büchi Labortechnik AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z563625-1EA | 4061832771496 |