Documenti fondamentali
Z680567
Caps for Duran® laboratory bottles
neck thread for GL 14, red PBT cap (with lip seal)
Sinonimo/i:
caps bottles, glass bottles caps
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
red PBT cap (with lip seal)
Produttore/marchio commerciale
DURAN®
Schott
Raccordi
neck thread for GL 14
Descrizione generale
The GL 14 caps and tubing inserts allow the DURAN® GL 45 2/3 port tubing connection caps or GL 45 / GLS 80® Stirred Bottle Reactors to be used with four different rigid or semi-rigid tubing outer diameters (1.6, 3.0, 3.2, or 6.0 mm). The different sized inserts form a tight seal around the pass-through tubing when used with the blue GL 14 tubing cap. Sterile pressure equalisation is possible through use of 0.2 µm syringe filter set. Unused ports can be sealed off with a red PBT GL 14 screw cap.
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
Note legali
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)