Documenti fondamentali
Z680400
Duran® GL 45 connection system caps and accessories
GL 45 blue PP screw cap with 3 x GL 14 ports
Sinonimo/i:
1129751, Connection System Accessories, Duran Connection Caps, GL 45 Connection System
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
blue-gray polypropylene
Produttore/marchio commerciale
DURAN® 1129751
Parametri
-40-140 °C temp. limit
Descrizione generale
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available: 1.6 mm, 3.0 mm, 3.2 mm and 6.0 mm that can be connected.
Sterile pressure equalization is possible through use of membrane filter.
Caratteristiche e vantaggi
  • Manufactured from Polypropylene._x000D_
  • Rigid or semi-rigid tubing of different diameters can be accepted by using a blue GL 14 open topped cap with one of the inserts._x000D_
  • Available with 2, or 3 threaded outlets (ports)._x000D_
  • Four different tubing diameters (1.6 mm, 3.0 mm, 3.2 mm or 6.0 mm tubing outer diameter) can be used by changing the GL 14 inserts._x000D_
  • By applying positive or negative pressure liquid can be pumped out of or into the bottle via the screw cap._x000D_
  • Fully compatible with all the GL 45 threads and therefore highly versatile.
Note legali
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)