Z558095
Spinner turbines for Bruker spectrometers
variable temperature, PET, diam. 5 mm
Sinonimo/i:
Wilmad nmr tubes, nmr sample tubes, nmr tubes
About This Item
Codice UNSPSC:
41121705
NACRES:
SB.32
Prodotti consigliati
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Wilmad B-PEEK-5
Diametro
5 mm
Descrizione generale
Fiberglass turbines are made from high density GFK fiberglass and incorporate a single O-ring imbedded at the turbine top to hold the sample tube firmly into place. The PET spinner turbine for Bruker spectrometers incorporates two O-rings for ease of use and snug fit on the sample tube. The Bruker PET spinner turbine is recommended for 500 and 600 MHz NMR spectrometers.
VT operation of -150°C to +200°C
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z558095-1EA | 4061832803289 |