Z303232
Quickfit® Liebig condenser
L 400 mm, joint: ST/NS 29/32
Sinonimo/i:
C1/44/SC
About This Item
Prodotti consigliati
Giunto
joint: ST/NS29/32
Produttore/marchio commerciale
Quickfit C1/44/SC
Disponibilità
not available in USA and Canada
not available in USA and Canada
Lunghezza
400 mm
Area superficiale
290 cm2
Descrizione generale
Designed to give a turbulent flow in the water jacket for efficient cooling at a comparatively slow rate of water flow. These condensers are adequate and effective in many applications.
Note legali
Quickfit is a registered trademark of SciLabware Limited