BR41638
BRAND® graduated cylinder low form embossed scale
volume 100 mL, polypropylene
Sinonimo/i:
Graduated Cylinders, Plastics
About This Item
Codice UNSPSC:
41121805
NACRES:
NB.43
Prodotti consigliati
Materiali
polypropylene
Produttore/marchio commerciale
BRAND 41638
Volume
100 mL
applicazioni
PFAS testing
Categorie correlate
Descrizione generale
The BRAND® Graduated Cylinder, Low Form with Embossed Scale, is designed for precise liquid measurement in laboratory settings. Made from high-clarity polypropylene (PP), this cylinder is calibrated to contain (TC, In), ensuring accurate volume readings. Its low form design enhances stability and ease of use, while the embossed scale provides clear visibility for quick reference. This graduated cylinder can withstand exposure to temperatures up to 80 °C without exceeding permanent error limits, making it suitable for a variety of applications. Reliable and durable, this graduated cylinder is an essential tool for any laboratory.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR41638-10EA | 4061831856354 |