BR41662
BRAND® graduated cylinder low form embossed scale
volume 1000 mL, polypropylene
Sinonimo/i:
Graduated Cylinders, Plastics
About This Item
Codice UNSPSC:
41121805
NACRES:
NB.43
Prodotti consigliati
Materiali
polypropylene
Produttore/marchio commerciale
BRAND 41662
Volume
1000 mL
Descrizione generale
The BRAND® Graduated Cylinder, Low Form with Embossed Scale, is designed for precise liquid measurement in laboratory settings. Made from high-clarity polypropylene (PP), this cylinder is calibrated to contain (TC, In), ensuring accurate volume readings. Its low form design enhances stability and ease of use, while the embossed scale provides clear visibility for quick reference. This graduated cylinder can withstand exposure to temperatures up to 80 °C without exceeding permanent error limits, making it suitable for a variety of applications. Reliable and durable, this graduated cylinder is an essential tool for any laboratory.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)