Z268658
Scienceware® Safe-Lab™ PTFE stopper
joint: ST/NS 45/50, hollow
Sinonimo/i:
lab stopper, lab stoppers, stoppers
About This Item
Codice UNSPSC:
41122500
NACRES:
NB.35
Prodotti consigliati
Giunto
joint: ST/NS45/50
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Bel-Art F20197-1550
Descrizione generale
PTFE stoppers with PP extracting nut for easy removal from flask. For use with boiling and distilling flasks with ground joints.
Note legali
Safe-Lab is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z268658-1EA | 4061832262888 |