Z693847
Scienceware® Break-Safe™ ampule opener
allows one handed opening of up to 3 ampules at a time
Sinonimo/i:
ampule breakers, glass ampoule openers
About This Item
Codice UNSPSC:
41121807
NACRES:
NB.43
Prodotti consigliati
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F38952-0000
Lungh. × Largh. × Alt.
9.5 cm (3.75 in.) × 7.6 cm (3 in.) × 8.3 cm (3.25 in.)
Descrizione generale
Unique, box style design allows one handed opening of up to 3 ampules at a time by simply pressing down on lid. Ampule necks snap clean and are contained within the device while fingers stay away from sharp edges.
After removing ampule contents, turn the device upside down over a trash receptacle to safely discard ampules and tops.
After removing ampule contents, turn the device upside down over a trash receptacle to safely discard ampules and tops.
- Includes 4 inserts to allow opening of 1 mL, 2 mL, 5 mL or 10 mL ampules
- Acrylic copolymer plastic construction has broad chemical resistance
- Transparency allows unobstructed viewing of ampule contents at all times
Note legali
Break-Safe is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)