Z225622
Stainless steel forceps
Specimen, L 12 in.
About This Item
Codice UNSPSC:
41122405
NACRES:
NB.35
Prodotti consigliati
Confezionamento
pack of 1 ea
Lunghezza
12 in.
Descrizione generale
with serrated tips
Applicazioni
Stainless steel forceps have been used for specimen embedding.
Caratteristiche e vantaggi
- Available in different sizes, styles, and designs
- Tips: pointed, blunt, rounded, curved, angled, or flat
- Inner surfaces: plain or serrated for extra grip
- Defined dimensions for precision work
- Foam or silicone-covered tips for extra grip
- Self-closing/locking guide pins to keep the tips aligned for a firmer grip
- Inert polytetrafluoroethylene (PTFE) coating prevents contamination, scratching, or damage to delicate samples
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)