Documenti fondamentali
Z168734
Stainless steel forceps
Specimen, L 6 in.
About This Item
Codice UNSPSC:
41122405
NACRES:
NB.35
Confezionamento
pack of 1 ea
Lunghezza
6 in.
Descrizione generale
with serrated tips
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 10
1 of 10
Stainless steel forceps Dissecting
Z168793
Stainless steel forceps
Dressing tissue forceps L 12 in.
F4642
Dressing tissue forceps
Stainless steel forceps Dumont
Z168742
Stainless steel forceps
Dressing tissue forceps L 8 in.
F4392
Dressing tissue forceps
Stainless steel forceps Fine tip, curved
Z168785
Stainless steel forceps
Tweezers Style #5, Fine needle-sharp, Inox alloy (not anti-magnetic)
T4662
Tweezers
Stainless steel forceps Fine tip, straight
Z168777
Stainless steel forceps
Micro-dissecting forceps straight
F4017
Micro-dissecting forceps
Stainless steel forceps High-precision
Z225304
Stainless steel forceps
Global Trade Item Number
SKUGTIN
Z168734-1EA4061833298879