Documenti fondamentali
Z223360
Sigma-Aldrich
Aldrich® three-way adapter with thermometer joint
joint: ST/NS 24/40
Sinonimo/i:
adapter laboratory, glass adapter, glass adapters
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Giunto
thermometer joint ST/NS10/30
joint: ST/NS24/40
Descrizione generale
Side-arm is at 75° angle from bottom joint. Distance from joint to side-arm is 75mm.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)