Z210544
Pro-Pak® distillation packing
0.16 in.2, Monel
About This Item
Codice UNSPSC:
41104812
NACRES:
NB.43
Prodotti consigliati
Confezionamento
pkg of 1 L
Produttore/marchio commerciale
Ace Glass 662409
Descrizione generale
Efficient, self-wetting, protruded metal dump-packing for distillation columns of 0.75 to 12 in. diameter. Maximum reflux rate of approximately 3.4 L/h. Free space for 0.16 in.2 size is 94% and for 0.24 in.2 size is 96%. Each package has a packing volume of approx. 1 L. Packed heights up to 20 ft possible without distortion.
Note legali
Pro-Pak is a registered trademark of Cannon Instruments Co.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z210544-1PAK | 4061832348599 |