Z166146
Aldrich® antisplash splash-guard adapter with return hole
female joint: ST/NS 24/40, male joint: ST/NS 24/40, capacity 500 mL
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Prodotti consigliati
Giunto
female joint: ST/NS24/40
male joint: ST/NS24/40
Capacità
500 mL
Categorie correlate
Descrizione generale
These traps greatly reduce carry-over from foaming or bumping. Anything that may climb can return back down through the return hole at the bottom of this adapter.
Applicazioni
Aldrich antisplash splash-guard adapter with return hole may be used between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into the condenser in case of "bumping". A drain hole at the base of the inner tube allows solvent to drain back into the evaporation flask.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z166146-1EA | 4061837608735 |