Z147796
Aldrich® antisplash splash-guard adapter without return hole
female joint: ST/NS 24/40, male joint: ST/NS 24/40, capacity 250 mL
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Prodotti consigliati
Giunto
female joint: ST/NS24/40
male joint: ST/NS24/40
Capacità
250 mL
Categorie correlate
Descrizione generale
Aldrich® antisplash splash-guard adapter without return hole are traps that greatly reduce carry-over from foaming or bumping. Whatever comes up into this trap will stay and not go back to the evaporator flask.
Applicazioni
Aldrich® antisplash splash-guard adapter without return hole is ideal for use between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into condenser in case of "bumping".
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z147796-1EA | 4061837603471 |