Documenti fondamentali
Z146544
Sigma-Aldrich
Ace Rotary-Evaporator/Freeze-Drying Flask Adapter Head W/ O-ring
joint: ST/NS 24/40
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Giunto
joint: ST/NS24/40
Descrizione
with FETFE®
O-ring.
Produttore/marchio commerciale
Ace Glass 703021
Descrizione generale
Flange with two ground flats opposite each other allows nylon retainers on adapter head to pass over. O-ring in head and flask grooves make a vacuum-tight seal when head is turned 90° in either direction. No grease, springs, or clamp necessary. Flasks sold separately.
Note legali
FETFE is a registered trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)